missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(3-Methylsulfonylphenyl)-1-propylpiperidine, TRC
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Specifications
Specifications
| Chemical Name or Material | 4-[3-(Methylsulfonyl)phenyl]-1-propylpiperidine |
| CAS | 346688-38-8 |
| Molecular Formula | C15 H23 N O2 S |
| Quantity | 50 mg |
| Synonym | Pridopidine |
| InChI Formula | InChI=1S/C15H23NO2S/c1-3-9-16-10-7-13(8-11-16)14-5-4-6-15(12-14)19(2,17)18/h4-6,12-13H,3,7-11H2,1-2H3 |
| SMILES | CCCN1CCC(CC1)c2cccc(c2)S(=O)(=O)C |
| IUPAC Name | 4-(3-methylsulfonylphenyl)-1-propylpiperidine |
| Molecular Weight (g/mol) | 281.41 |
| Formula Weight | 281.1449 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction