missing translation for 'onlineSavingsMsg'
Learn More

4-[4-(4-Chlorophenyl)-4-hydroxypiperidin-1-yl]-1-[4-[4-(4-chlorophenyl)-4-hydroxypiperidin-1-yl]phenyl]butan-1-one, Mikromol™

Product Code. 30049272
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30049272 25 mg 25mg
17932186 100 mg 100mg
30459878 4 x 25 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30049272 Supplier Mikromol Supplier No. MM1165.040025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

4-[4-(4-Chlorophenyl)-4-hydroxypiperidin-1-yl]-1-[4-[4-(4-chlorophenyl)-4-hydroxypiperidin-1-yl]phenyl]butan-1-one
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4,4'-Bis[4-(p-chlorophenyl)-4-hydroxypiperidino]-butyrophenone
CAS 67987-08-0
Analyte or Component Names 4,4'-Bis[4-(p-chlorophenyl)-4-hydroxypiperidino]-butyrophenone
Shipping Condition Room Temperature
Molecular Formula C32 H36 Cl2 N2 O3
Certifications/Compliance ISO 9001
Synonym Haloperidol Imp. D (EP), Haloperidol USP Related Compound A, Haloperidol USP RC A, 1-Butanone, 4-[4-(4-chlorophenyl)-4-hydroxy-1-piperidinyl]-1-[4-[4-(4-chlorophenyl)-4-hydroxy-1-piperidinyl]phenyl]-, Haloperidol Related Compound A (USP), Haloperidol Related Compound A, Haloperidol Impurity D
InChI Formula InChI=1S/C32H36Cl2N2O3/c33-27-9-5-25(6-10-27)31(38)15-20-35(21-16-31)19-1-2-30(37)24-3-13-29(14-4-24)36-22-17-32(39,18-23-36)26-7-11-28(34)12-8-26/h3-14,38-39H,1-2,15-23H2
SMILES OC1(CCN(CCCC(=O)c2ccc(cc2)N3CCC(O)(CC3)c4ccc(Cl)cc4)CC1)c5ccc(Cl)cc5
IUPAC Name 4-[4-(4-chlorophenyl)-4-hydroxypiperidin-1-yl]-1-[4-[4-(4-chlorophenyl)-4-hydroxypiperidin-1-yl]phenyl]butan-1-one
Molecular Weight (g/mol) 567.55
Quantity 25 mg
Formula Weight 566.2103 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.