Learn More
4-(4-Chlorophenyl)-2-butanone, TRC
Specifications
Specifications
| Chemical Name or Material | 4-(4-Chlorophenyl)-2-butanone |
| CAS | 3506-75-0 |
| Molecular Formula | C10H11ClO |
| Quantity | 10 g |
| Synonym | 4-(p-Chlorophenyl)-2-butanone, NSC 348, p-Chlorobenzylacetone, 4-(4-Chlorophenyl)butan-2-one |
| InChI Formula | InChI=1S/C10H11ClO/c1-8(12)2-3-9-4-6-10(11)7-5-9/h4-7H,2-3H2,1H3 |
| SMILES | ClC1=CC=C(CCC(C)=O)C=C1 |
| IUPAC Name | 4-(4-chlorophenyl)butan-2-one |
| Molecular Weight (g/mol) | 182.65 |
| Formula Weight | 182.05 |