missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Dibromotriphenylamine, 98%, Thermo Scientific Chemicals

Catalog Number 15411969
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
81090-53-1
C18H13Br2N
403.117
MFCD00060104
KIGVOJUDEQXKII-UHFFFAOYSA-N
4,4'-dibromotriphenylamine, 4-bromo-n-4-bromophenyl-n-phenylaniline, n,n-bis 4-bromophenyl aniline, 4,4-dibromotriphenylamine, 4-bromo-n-4-bromophenyl-n-phenylbenzenamine, dibromotriphenylamine, 4,4'-dibromotriphenylamine;, 4,4'-dibromo-triphenylamine, bis 4-bromophenyl phenylamine, bis 4-bromophenyl phenylamine;
13594707
4-bromo-N-(4-bromophenyl)-N-phenylaniline
C1=CC=C(C=C1)N(C2=CC=C(C=C2)Br)C3=CC=C(C=C3)Br
This item is not returnable. View return policy
81090-53-1
C18H13Br2N
403.117
MFCD00060104
KIGVOJUDEQXKII-UHFFFAOYSA-N
4,4'-dibromotriphenylamine, 4-bromo-n-4-bromophenyl-n-phenylaniline, n,n-bis 4-bromophenyl aniline, 4,4-dibromotriphenylamine, 4-bromo-n-4-bromophenyl-n-phenylbenzenamine, dibromotriphenylamine, 4,4'-dibromotriphenylamine;, 4,4'-dibromo-triphenylamine, bis 4-bromophenyl phenylamine, bis 4-bromophenyl phenylamine;
13594707
4-bromo-N-(4-bromophenyl)-N-phenylaniline
C1=CC=C(C=C1)N(C2=CC=C(C=C2)Br)C3=CC=C(C=C3)Br

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 85°C
Color White
Flash Point >110°C (230°F)
Quantity 250 mg
Beilstein 7094354
Formula Weight 403.12
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 4,4'-Dibromotriphenylamine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'