Learn More
4,4-Dimethylcyclohexanone, 98%, Thermo Scientific Chemicals
Product Code. 11455600
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11455600 1 g 1g
15413327 250 mg 250mg
11465600 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11455600 Supplier Thermo Scientific Alfa Aesar Supplier No. H25857.03
 Restricted Item
Estimated Shipment: 05-10-2026
Add to basket
4,4-Dimethylcyclohexanone is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4,4-Dimethylcyclohexanone is used as a pharmaceutical intermediate.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible material is strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 4255-62-3
Molecular Formula C8H14O
Molecular Weight (g/mol) 126.199
MDL Number MFCD00234965
InChI Key PXQMSTLNSHMSJB-UHFFFAOYSA-N
Synonym 4,4-dimethylcyclohexanone, cyclohexanone, 4,4-dimethyl, 4,4-dimethyl cyclohexanone, 4,4-dimethyl-cyclohexan-1-one, 4,4-dimethycyclohexanone, 4.4-dimethylcyclohexanone, acmc-209jq8, 4,4-dimethyl-cyclohexanone, ksc494a0f, 4,4-dimethyl-1-cyclohexanone
PubChem CID 138166
IUPAC Name 4,4-dimethylcyclohexan-1-one
SMILES CC1(CCC(=O)CC1)C
Specifications
Melting Point 41°C to 43°C
Flash Point 62°C (144°F)
Quantity 1 g
Solubility Information Insoluble in water.
Formula Weight 126.2
Percent Purity 98%
Chemical Name or Material 4,4-Dimethylcyclohexanone