missing translation for 'onlineSavingsMsg'
Learn More

[4-(4-Hydroxy-3,5-diiodophenoxy)-3,5-diiodophenyl]acetic Acid (Tetraiodothyroacetic Acid), Mikromol™

Product Code. 30051190
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
This item is not returnable. View return policy

Product Code. 30051190

Brand: Mikromol MM0626.050025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

[4-(4-Hydroxy-3,5-diiodophenoxy)-3,5-diiodophenyl]acetic Acid (Tetraiodothyroacetic Acid)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3,5,3',5'-Tetraiodo Thyroacetic Acid
CAS 67-30-1
Analyte or Component Names 3,5,3',5'-Tetraiodo Thyroacetic Acid
Shipping Condition Room Temperature
Molecular Formula C14 H8 I4 O4
Certifications/Compliance ISO 9001
Synonym Liothyronine Sodium Imp. D (EP), Liothyronine Imp. D (EP), Benzeneacetic acid, 4-(4-hydroxy-3,5-diiodophenoxy)-3,5-diiodo-, Acetic acid, [4-(4-hydroxy-3,5-diiodophenoxy)-3,5-diiodophenyl]- (8CI), Thyroacetic acid, 3,3',5,5'-tetraiodo- (6CI), 3,3',5,5'-Tetraiodothyroacetic acid, 3,5,3',5'-Tetraiodothyroacetic acid, Tetrac, Tetraiodothyroacetic acid, Levothyroxine Sodium Impurity D, Liothyronine Sodium Impurity D, Levothyroxine Impurity D, Liothyronine Impurity D
InChI Formula InChI=1S/C14H8I4O4/c15-8-4-7(5-9(16)13(8)21)22-14-10(17)1-6(2-11(14)18)3-12(19)20/h1-2,4-5,21H,3H2,(H,19,20)
SMILES OC(=O)Cc1cc(I)c(Oc2cc(I)c(O)c(I)c2)c(I)c1
IUPAC Name 2-[4-(4-hydroxy-3,5-diiodophenoxy)-3,5-diiodophenyl]acetic acid
Molecular Weight (g/mol) 747.83
Quantity 25 mg
Formula Weight 747.6601 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.