Learn More
4,5-Diamino-6-chloropyrimidine, TRC
Specifications
Specifications
| Chemical Name or Material | 4,5-Diamino-6-chloropyrimidine |
| CAS | 4316-98-7 |
| Molecular Formula | C4 H5 Cl N4 |
| Quantity | 2.5 g |
| Synonym | 4-Chloropyrimidine-5,6-diamine, 5,6-Diamino-4-chloropyrimidine, 6-Chloropyrimidine-4,5-diamine, NSC 36907, 6-Chloro-4,5-pyrimidinediamine |
| InChI Formula | InChI=1S/C4H5ClN4/c5-3-2(6)4(7)9-1-8-3/h1H,6H2,(H2,7,8,9) |
| SMILES | Nc1ncnc(Cl)c1N |
| IUPAC Name | 6-chloropyrimidine-4,5-diamine |
| Molecular Weight (g/mol) | 144.562 |
| Formula Weight | 144.02 |