Learn More
4,5-Difluoro-2-methoxybenzyl bromide, 97%, Thermo Scientific™
Product Code. 11336839
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11336839 1 g 1g
11346839 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11336839 Supplier Thermo Scientific Alfa Aesar Supplier No. H26764.03
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 886499-64-5
Molecular Formula C8H7BrF2O
Molecular Weight (g/mol) 237.044
MDL Number MFCD06660220
InChI Key PRLKVWCFDNQAED-UHFFFAOYSA-N
Synonym 4,5-difluoro-2-methoxybenzyl bromide, 1-bromomethyl-4,5-difluoro-2-methoxybenzene
PubChem CID 17750791
IUPAC Name 1-(bromomethyl)-4,5-difluoro-2-methoxybenzene
SMILES COC1=CC(=C(C=C1CBr)F)F
Specifications
Melting Point 40°C to 43°C
Quantity 1 g
UN Number UN3261
Formula Weight 237.04
Percent Purity 97%
Chemical Name or Material 4,5-Difluoro-2-methoxybenzyl bromide
Name des Produkts
Wählen Sie ein Thema
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.