Learn More
4,5-Dimethoxy-2-nitrobenzyl carbonochloridate, 97%, BLD Pharm™
Specifications
Specifications
| CAS | 42855-00-5 |
| Molecular Formula | C10H10ClNO6 |
| MDL Number | MFCD00143507 |
| Quantity | 100 mg |
| SMILES | O=C(Cl)OCC1=CC(OC)=C(OC)C=C1[N+]([O-])=O |
| Molecular Weight (g/mol) | 275.64 |
| Percent Purity | 97% |
| Chemical Name or Material | 4,5-Dimethoxy-2-nitrobenzyl carbonochloridate |