missing translation for 'onlineSavingsMsg'
Learn More

4,5-Dimethyl-o-phenylenediamine, 98%, many contain up to ca 15% water

Product Code. p-7031571
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 15468165

Brand: Thermo Scientific Alfa Aesar H32695.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3171-45-7 | C8H12N2 | 136.20 g/mol

4, 5-dimethyl-o-phenylenediamine used in the determination of aromatic aldehydes by high performance liquid chromatography after precolumn fluorescent derivatization. For the determination of vanillin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4, 5-dimethyl-o-phenylenediamine used in the determination of aromatic aldehydes by high performance liquid chromatography after precolumn fluorescent derivatization. For the determination of vanillin.

Solubility
Slightly soluble in water.

Notes
Air & Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air, heat, light, acid chlorides, acid anhydrides.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3171-45-7
Molecular Formula C8H12N2
Molecular Weight (g/mol) 136.20
MDL Number MFCD00007729
InChI Key XSZYBMMYQCYIPC-UHFFFAOYSA-N
Synonym 4,5-dimethyl-1,2-phenylenediamine, 4,5-diamino-o-xylene, 4,5-dimethyl-o-phenylenediamine, 1,2-benzenediamine, 4,5-dimethyl, 1,2-diamino-4,5-dimethylbenzene, 4,5-dimethyl-1,2-benzenediamine, 4,5-dimethyl-benzene-1,2-diamine, 4,5-dimethyl-ortho-phenylenediamine, 4,5-dimethylphenylene-1,2-diamine, 4,5-dimethyl-o-phenylenediamine monohydrate
PubChem CID 76635
IUPAC Name 4,5-dimethylbenzene-1,2-diamine
SMILES CC1=CC(N)=C(N)C=C1C

Specifications

Melting Point 127°C to 129°C
Quantity 5 g
UN Number UN2811
Solubility Information Slightly soluble in water.
Formula Weight 136.2
Percent Purity 98%
Chemical Name or Material 4,5-Dimethyl-o-phenylenediamine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.