Learn More
4,5-Diphenylimidazolin-2-one, 98%, Thermo Scientific™
Product Code. 11341867
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11341867 5 g 5g
11331867 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11341867 Supplier Thermo Scientific Alfa Aesar Supplier No. L07340.06
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 642-36-4
Molecular Formula C15H12N2O
MDL Number MFCD00022393
Synonym 4,5-Diphenyl-2-glyoxalone
Specifications
Melting Point 323°C to 326°C
Quantity 5 g
Beilstein 187611
Formula Weight 236.28
Percent Purity 98%
Chemical Name or Material 4,5-Diphenylimidazolin-2-one