Learn More
4-[5-(Trifluoromethyl)pyrid-2-yl]benzoic acid, 95%, Thermo Scientific™
Product Code. 10411324
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10411324 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10411324 Supplier Thermo Scientific Maybridge Supplier No. CC53001DA
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket
Specifications
CAS 223127-47-7
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Molecular Formula C13H8F3NO2
MDL Number MFCD09817475
Quantity 1 g
Synonym 4-5-trifluoromethyl pyridin-2-yl benzoic acid, 4-5-trifluoromethyl pyrid-2-yl benzoic acid, benzoic acid,4-5-trifluoromethyl-2-pyridinyl, 4-5-trifluoromethylpyridin-2-yl benzoic acid, 4-5-trifluoromethyl-2-pyridyl benzenecarboxylic acid
InChI Key FSLUYPHZCSKJJZ-UHFFFAOYSA-N
SMILES OC(=O)C1=CC=C(C=C1)C1=NC=C(C=C1)C(F)(F)F
IUPAC Name 4-[5-(trifluoromethyl)pyridin-2-yl]benzoic acid
Molecular Weight (g/mol) 267.21
PubChem CID 22730671
Formula Weight 267.21
Percent Purity 95%
Chemical Name or Material 4-[5-(Trifluoromethyl)pyrid-2-yl]benzoic acid