Learn More
4,6-Dihydroxy-5-(2-methoxyphenoxy)-2-(pyrimidin-2-yl)pyrimidine, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
4,6-Dihydroxy-5-(2-methoxyphenoxy)-2-(pyrimidin-2-yl)pyrimidine
Specifications
Specifications
| Chemical Name or Material | 4,6-Dihydroxy-5-(o-methoxyphenoxy)-2,2'-bipyrimidine |
| CAS | 329923-15-1 |
| Analyte or Component Names | 4,6-Dihydroxy-5-(o-methoxyphenoxy)-2,2'-bipyrimidine |
| Shipping Condition | Room Temperature |
| Molecular Formula | C15 H12 N4 O4 |
| Certifications/Compliance | ISO 17025 |
| Synonym | [2,2'-Bipyrimidin]-4(3H)-one, 6-hydroxy-5-(2-methoxyphenoxy)-, [2,2'-Bipyrimidin]-4(1H)-one, 6-hydroxy-5-(2-methoxyphenoxy)- (9CI), 4,6-Dihydroxy-5-(o-methoxyphenoxy)-2-(2-pyrimidinyl)pyrimidine, 4,6-Dihydroxy-5-(2-methoxyphenoxy)-2-(pyrimidin-2-yl)pyrimidine |
| InChI Formula | InChI=1S/C15H12N4O4/c1-22-9-5-2-3-6-10(9)23-11-14(20)18-13(19-15(11)21)12-16-7-4-8-17-12/h2-8H,1H3,(H2,18,19,20,21) |
| SMILES | COc1ccccc1Oc2c(O)nc(nc2O)c3ncccn3 |
| IUPAC Name | 5-(2-methoxyphenoxy)-2-pyrimidin-2-ylpyrimidine-4,6-diol |
| Show More |