Learn More
4-Acetoxybenzaldehdye, TRC
Specifications
Specifications
| Chemical Name or Material | 4-Acetoxybenzaldehdye |
| CAS | 878-00-2 |
| Molecular Formula | C9H8O3 |
| Quantity | 100 mg |
| Synonym | 4-Acetoxybenzaldehyde |
| InChI Formula | InChI=1S/C9H8O3/c1-7(11)12-9-4-2-8(6-10)3-5-9/h2-6H,1H3 |
| SMILES | CC(=O)Oc1ccc(C=O)cc1 |
| IUPAC Name | (4-formylphenyl) acetate |
| Molecular Weight (g/mol) | 164.16 |
| Formula Weight | 164.0473 |