missing translation for 'onlineSavingsMsg'
Learn More

4-Allylanisole, 98%, Thermo Scientific Chemicals

Catalog Number p-51841
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
140-67-0
C10H12O
148.2
MFCD00008653
ZFMSMUAANRJZFM-UHFFFAOYSA-N
estragole, 4-allylanisole, p-allylanisole, tarragon, methyl chavicol, 1-allyl-4-methoxybenzene, estragol, esdragol, esdragole, esdragon
8815
CHEBI:4867
1-methoxy-4-prop-2-enylbenzene
COC1=CC=C(C=C1)CC=C
This item is not returnable. View return policy
140-67-0
C10H12O
148.2
MFCD00008653
ZFMSMUAANRJZFM-UHFFFAOYSA-N
estragole, 4-allylanisole, p-allylanisole, tarragon, methyl chavicol, 1-allyl-4-methoxybenzene, estragol, esdragol, esdragole, esdragon
8815
CHEBI:4867
1-methoxy-4-prop-2-enylbenzene
COC1=CC=C(C=C1)CC=C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Color Colorless
Density 0.9600g/mL
Boiling Point 215°C to 216°C
Flash Point 81°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula H2C=CHCH2C6H4OCH3
Refractive Index 1.5205 to 1.5225
Quantity 5 g
Beilstein 06, 571
Merck Index 15, 3760
Specific Gravity 0.96
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol and chloroform
Formula Weight 148.2
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 4-Allylanisole, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Allylanisole
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'