missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-1-(3-chloro-2-pyridyl)piperidine hydrochloride, 97%, Thermo Scientific™

Product Code. 15449425
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15449425 250 mg 250mg
15439425 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15449425 Supplier Thermo Scientific Alfa Aesar Supplier No. H51927.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77145-35-8
Molecular Formula C10H15Cl2N3
Molecular Weight (g/mol) 248.15
MDL Number MFCD16251586
InChI Key NOSKSNWYGYGGBH-UHFFFAOYSA-N
Synonym 4-amino-1-3-chloro-2-pyridyl piperidine hydrochloride, 1-3-chloropyridin-2-yl piperidin-4-amine hydrochloride, 1-3-chloro-2-pyridyl piperidin-4-amine hydrochloride, 1-3-chloro-2-pyridinyl-4-piperidinamine hydrochloride 1:1
PubChem CID 69359259
IUPAC Name 1-(3-chloropyridin-2-yl)piperidin-4-amine;hydrochloride
SMILES Cl.NC1CCN(CC1)C1=C(Cl)C=CC=N1

Specifications

Melting Point 265°C to 267°C
Quantity 250 mg
Formula Weight 248.15
Percent Purity 97%
Chemical Name or Material 4-Amino-1-(3-chloro-2-pyridyl)piperidine hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.