Learn More
4-Amino-2-methyl-1H-isoindole-1,3(2H)-dione-13C4, TRC
Product Code. 30328466
Change view
Click to view available options
Quantity:
0.5 mg
5 mg
Unit Size:
0.5mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30328466 5 mg 5mg
30313747 0.5 mg 0.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30328466 Supplier Toronto Research Chemicals Supplier No. TRCA6170625MG
 Restricted Item
Estimated Shipment: 28-07-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 4-Amino-2-methyl-1H-isoindole-1,3(2H)-dione-13C4
Molecular Formula C513C4H8N2O2
Quantity 5 mg
Synonym 4-Amino-2-methyl-1H-isoindole-1,3(2H)-dione-13C4, 3-Amino-N-methylphthalimide-13C4
InChI Formula InChI=1S/C9H8N2O2/c1-11-8(12)5-3-2-4-6(10)7(5)9(11)13/h2-4H,10H2,1H3/i2+1,3+1,4+1,6+1
SMILES O=C1N(C)C(C2=[13CH][13CH]=[13CH][13C](N)=C21)=O
IUPAC Name 4-amino-2-methylisoindoline-1,3-dione-4,5,6,7-13C4
Molecular Weight (g/mol) 180.14
Formula Weight 180.07