Learn More
4-Amino-3,5-dichlorobenzonitrile, 97%, Thermo Scientific™
Product Code. 11443624
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11443624 25 g 25g
11423624 1 g 1g
11433624 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11443624 Supplier Thermo Scientific Alfa Aesar Supplier No. A11269.14
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Voir les produits de remplacement
missing translation for 'validMainframeMsg'
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Identifiants chimiques
CAS 78473-00-4
Molecular Formula C7H4Cl2N2
Molecular Weight (g/mol) 187.023
MDL Number MFCD00017341
InChI Key COFNCCWGWXFACE-UHFFFAOYSA-N
Synonym 2,6-dichloro-4-cyanoaniline, 4-cyano-2,6-dichloroaniline, benzonitrile, 4-amino-3,5-dichloro, 3,5-dichloro-4-aminobenzonitrile, 4-amino-3,5-dichloro-benzonitrile, 4-amino-3,5-dichlorobenzenecarbonitrile, pubchem14453, acmc-1be7s, ksc381a4h, timtec-bb sbb003696
PubChem CID 522706
IUPAC Name 4-amino-3,5-dichlorobenzonitrile
SMILES C1=C(C=C(C(=C1Cl)N)Cl)C#N
Spécification
Melting Point 117°C to 119°C
Quantity 25 g
UN Number UN3439
Beilstein 2719605
Formula Weight 187.03
Percent Purity 97%
Chemical Name or Material 4-Amino-3,5-dichlorobenzonitrile