missing translation for 'onlineSavingsMsg'
Learn More

4-Aminoresorcinol Hydrochloride, TRC

Product Code. 30324365
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
missing translation for 'unitSize':
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity missing translation for 'unitSize'
30324365 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 30324365 missing translation for 'mfr' Toronto Research Chemicals missing translation for 'supplierNo' TRCA6292701G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Aminoresorcinol Hydrochloride
CAS 34781-86-7
Molecular Formula C6H7NO2. HCl
Quantity 1 g
Synonym 4-Amino-1,3-benzenediol Hydrochloride, 2,4-Dihydroxyaniline Hydrochloride, 4-Aminobenzene-1,3-diol Hydrochloride
InChI Formula InChI=1S/C6H7NO2.ClH/c7-5-2-1-4(8)3-6(5)9;/h1-3,8-9H,7H2;1H
SMILES Cl.Nc1ccc(O)cc1O
IUPAC Name 4-aminobenzene-1,3-diol;hydrochloride
Molecular Weight (g/mol) 161.58
Formula Weight 161.0244
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.