missing translation for 'onlineSavingsMsg'
Learn More

4-Benzyloxy Atorvastatin-d5 Acetonide tert-Butyl Ester, TRC

Product Code. 30334743
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30334743 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30334743 Supplier Toronto Research Chemicals Supplier No. TRCB2876521MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Benzyloxy Atorvastatin-d5 Acetonide tert-Butyl Ester
CAS 265989-37-5
Molecular Formula C47H48D5FN2O6
Quantity 1 mg
Synonym (4R,6R)-6-[2-[2-(4-fluorophenyl)-5-(1-methylethyl)-3-(phenyl-d5)-4-[[[4-(phenylmethoxy)phenyl]amino]carbonyl]-1H-pyrrol-1-yl]ethyl]-2,2-dimethyl-1,3-dioxane-4-acetic Acid 1,1-Dimethylethyl Ester, (6-{2-[3-(4-Benzyloxy-phenylcarbamoyl)-5-(4-fluoro-phenyl)-2-isopropyl-4-phenyl-d5-pyrrol-1-yl]-ethyl}-2,2-dimethyl-[1,3]-dioxane-4-yl)-acetic Acid, tert-Butyl Ester
InChI Formula InChI=1S/C47H53FN2O6/c1-31(2)43-42(45(52)49-36-22-24-37(25-23-36)53-30-32-14-10-8-11-15-32)41(33-16-12-9-13-17-33)44(34-18-20-35(48)21-19-34)50(43)27-26-38-28-39(55-47(6,7)54-38)29-40(51)56-46(3,4)5/h8-25,31,38-39H,26-30H2,1-7H3,(H,49,52)/i9D,12D,13D,16D,17D
SMILES [2H]c1c([2H])c([2H])c(c([2H])c1[2H])c2c(C(=O)Nc3ccc(OCc4ccccc4)cc3)c(C(C)C)n(CCC5CC(CC(=O)OC(C)(C)C)OC(C)(C)O5)c2c6ccc(F)cc6
IUPAC Name tert-butyl 2-[6-[2-[2-(4-fluorophenyl)-3-(2,3,4,5,6-pentadeuteriophenyl)-4-[(4-phenylmethoxyphenyl)carbamoyl]-5-propan-2-ylpyrrol-1-yl]ethyl]-2,2-dimethyl-1,3-dioxan-4-yl]acetate
Molecular Weight (g/mol) 765.96
Formula Weight 765.4201
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.