missing translation for 'onlineSavingsMsg'
Learn More

4-Bromophenetole, TRC

Product Code. 30335376
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30335376 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30335376 Supplier Toronto Research Chemicals Supplier No. TRCB686150100G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Bromophenetole
CAS 588-96-5
Molecular Formula C8H9BrO
Quantity 100 g
Synonym 1-Bromo-4-ethoxy-benzene, p-Bromo-phenetole, 1-Bromo-4-ethoxybenzene, 4-Bromoethoxybenzene, 4-Bromophenyl Ethyl Ether, 4-Ethoxybromobenzene, 4-Ethoxyphenyl Bromide, Ethyl 4-Bromophenyl Ether, NSC 8053, p-Bromoethoxybenzene, p-Bromophenetole, p-Bromophenol Ethyl Ether, p-Ethoxybromobenzene, p-Ethoxyphenyl Bromide
InChI Formula InChI=1S/C8H9BrO/c1-2-10-8-5-3-7(9)4-6-8/h3-6H,2H2,1H3
SMILES CCOc1ccc(Br)cc1
IUPAC Name 1-bromo-4-ethoxybenzene
Molecular Weight (g/mol) 201.06
Formula Weight 199.9837
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.