Learn More
4-Chloro-1-methoxy-2-methylbenzene, 95%, BLD Pharm™
Specifications
Specifications
| CAS | 3260-85-3 |
| Molecular Formula | C8H9ClO |
| MDL Number | MFCD00070771 |
| Quantity | 25 g |
| SMILES | CC1=CC(Cl)=CC=C1OC |
| Molecular Weight (g/mol) | 156.61 |
| Percent Purity | 95% |
| Chemical Name or Material | 4-Chloro-1-methoxy-2-methylbenzene |