missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2,5-difluorobenzoic acid, 98%, Thermo Scientific™

Product Code. 15451808
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15451808 5 g 5g
15461808 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15451808 Supplier Thermo Scientific Alfa Aesar Supplier No. H55521.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 132794-07-1
Molecular Formula C7H2ClF2O2
Molecular Weight (g/mol) 191.54
MDL Number MFCD00077480
InChI Key PEPCYJSDHYMIFN-UHFFFAOYSA-M
Synonym 2,5-difluoro-4-chlorobenzoic acid, 4-chloro-2,5-difluorobenzoicacid, 4-chloro-2,5-difluoro-benzoic acid, benzoic acid, 4-chloro-2,5-difluoro, pubchem1367, intermediates-zcf02018, acmc-1c2s0, 2,5-difluoro-4-chlorobenzoicacid, attercop-chm at136451, 2,5-difluoro-4-chloro-benzoic acid
PubChem CID 2733274
IUPAC Name 4-chloro-2,5-difluorobenzoic acid
SMILES [O-]C(=O)C1=CC(F)=C(Cl)C=C1F

Specifications

Melting Point 154°C to 157°C
Quantity 5 g
Formula Weight 192.55
Percent Purity 98%
Chemical Name or Material 4-Chloro-2,5-difluorobenzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.