missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2,6-difluorobenzaldehyde, Maybridge

Product Code. 10431990
Change view
Click to view available options
Quantity:
5 g
Packaging:
Amber Glass Bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10431990 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10431990 Supplier Thermo Scientific Maybridge Supplier No. MAY00225DE
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 252004-45-8
Packaging Amber Glass Bottle
Molecular Formula C7H3ClF2O
Quantity 5 g
Synonym benzaldehyde, 4-chloro-2,6-difluoro, pubchem8442, acmc-1chvl, 4-chloro-2,6-difluoro-benzaldehyde, benzaldehyde,4-chloro-2,6-difluoro, 4-chloro-2,6-difluorobenzaldehyde, 4-chloranyl-2,6-bis fluoranyl benzaldehyde
InChI Key GTTVSBCPMJQRSP-UHFFFAOYSA-N
SMILES C1=C(C=C(C(=C1F)C=O)F)Cl
IUPAC Name 4-chloro-2,6-difluorobenzaldehyde
Molecular Weight (g/mol) 176.547
PubChem CID 2773512
Formula Weight 176.55
Percent Purity 97%
Chemical Name or Material 4-Chloro-2, 6-difluorobenzaldehyde
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.