missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-fluorobenzyl Chloride, TRC

Product Code. 30359589
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30359589 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30359589 Supplier Toronto Research Chemicals Supplier No. TRCC3713351G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 87417-71-8
Molecular Formula C7 H5 Cl2 F
Quantity 1 g
Synonym 4-Chloro-1-(chloromethyl)-2-fluorobenzene
InChI Formula InChI=1S/C7H5Cl2F/c8-4-5-1-2-6(9)3-7(5)10/h1-3H,4H2
SMILES Fc1cc(Cl)ccc1CCl
IUPAC Name 4-chloro-1-(chloromethyl)-2-fluorobenzene
Molecular Weight (g/mol) 179.019
Formula Weight 177.975
Chemical Name or Material 4-Chloro-2-fluorobenzyl Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.