missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-methylaniline, 98%, Thermo Scientific Chemicals

Catalog Number p-7026301
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
7149-75-9
C7H8ClN
141.598
MFCD00066332
HIHCTGNZNHSZPP-UHFFFAOYSA-N
5-amino-2-chlorotoluene, benzenamine, 4-chloro-3-methyl, m-toluidine, 4-chloro, 3-methyl-4-chloroaniline, 4-chloro-m-toluidine, unii-6q2w6l7bwz, 6q2w6l7bwz, 4-chloro-3-methylphenylamine, 4-chloro-3-methyl-phenylamine, 4-chloro-3-methylphenyl amine
23536
4-chloro-3-methylaniline
CC1=C(C=CC(=C1)N)Cl
This item is not returnable. View return policy
7149-75-9
C7H8ClN
141.598
MFCD00066332
HIHCTGNZNHSZPP-UHFFFAOYSA-N
5-amino-2-chlorotoluene, benzenamine, 4-chloro-3-methyl, m-toluidine, 4-chloro, 3-methyl-4-chloroaniline, 4-chloro-m-toluidine, unii-6q2w6l7bwz, 6q2w6l7bwz, 4-chloro-3-methylphenylamine, 4-chloro-3-methyl-phenylamine, 4-chloro-3-methylphenyl amine
23536
4-chloro-3-methylaniline
CC1=C(C=CC(=C1)N)Cl

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 82°C to 85°C
Quantity 5 g
UN Number UN2239
Beilstein 636522
Formula Weight 141.6
Percent Purity 98%
Chemical Name or Material 4-Chloro-3-methylaniline

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'