missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-sulfobenzoic Acid, TRC

Product Code. 30355625
Change view
Click to view available options
Quantity:
25 mg
100 mg
250 mg
Unit Size:
100mg
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30355625 25 mg -
30360687 250 mg 250mg
30349244 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30355625 Supplier Toronto Research Chemicals Supplier No. TRCC38275025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 755698-62-5
Molecular Formula C7H5ClO5S
Molecular Weight (g/mol) 236.63
Synonym 4-Chloror-3-Sulfo-Benzoic Acid
IUPAC Name 4-chloro-3-sulfobenzoic acid
SMILES OC(=O)c1ccc(Cl)c(c1)S(=O)(=O)O

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C7H5ClO5S/c8-5-2-1-4(7(9)10)3-6(5)14(11,12)13/h1-3H,(H,9,10)(H,11,12,13)
Formula Weight 235.9546
Chemical Name or Material 4-Chloro-3-sulfobenzoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.