Learn More
4-Chloro-6-ethyl-5-fluoropyrimidine, 95%, Thermo Scientific™
Product Code. 15448238
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15448238 25 g 25g
15438238 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15448238 Supplier Thermo Scientific Alfa Aesar Supplier No. H61674.14
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 137234-74-3
Molecular Formula C6H6ClFN2
Molecular Weight (g/mol) 160.58
MDL Number MFCD07782087
InChI Key LKTGVRWVTAJGMS-UHFFFAOYSA-N
Synonym 4-chloro-6-ethyl-5-fluoro-pyrimidine, pyrimidine, 4-chloro-6-ethyl-5-fluoro, 4-chloro-5-fluoro-6-ethylpyrimidine, pubchem7037, acmc-1bzf7, ksc174s6p
PubChem CID 16743445
IUPAC Name 4-chloro-6-ethyl-5-fluoropyrimidine
SMILES CCC1=C(F)C(Cl)=NC=N1
Specifications
Density 1.286
Boiling Point 201°C to 211°C
Flash Point 81°C (178°F)
Refractive Index 1.496
Quantity 25 g
UN Number UN3267
Formula Weight 160.58
Percent Purity 95%
Chemical Name or Material 4-Chloro-6-ethyl-5-fluoropyrimidine