Learn More
4-Chloro-7-hydroxy-6-methoxy-3-quinolinecarbonitrile, TRC
Specifications
Specifications
| Chemical Name or Material | 4-Chloro-7-hydroxy-6-methoxy-3-quinolinecarbonitrile |
| CAS | 263149-10-6 |
| Molecular Formula | C11H7ClN2O2 |
| Quantity | 500 mg |
| Synonym | 4-Chloro-3-cyano-7-hydroxy-6-methoxyquinoline, 4-Chloro-7-hydroxy-6-methoxy-3-quinolinecarbonitrile |
| InChI Formula | InChI=1S/C11H7ClN2O2/c1-16-10-2-7-8(3-9(10)15)14-5-6(4-13)11(7)12/h2-3,5,15H,1H3 |
| SMILES | COc1cc2c(Cl)c(cnc2cc1O)C#N |
| IUPAC Name | 4-chloro-7-hydroxy-6-methoxyquinoline-3-carbonitrile |
| Molecular Weight (g/mol) | 234.64 |
| Formula Weight | 234.0196 |