missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-N-(2-methyl-1H-indol-1-yl)-3-sulphamoylbenzamide, Mikromol™

Product Code. 30049543
Click to view available options
Quantity:
25 mg
100 mg
missing translation for 'unitSize'
100mg
25mg
This item is not returnable. View return policy

Product Code. 30049543

missing translation for 'mfr': Mikromol MM0477.020025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

4-Chloro-N-(2-methyl-1H-indol-1-yl)-3-sulphamoylbenzamide
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-chloro-3-sulfamoyl-N-(2-methyl-1H-indol-1-yl)benzamide
CAS 63968-75-2
Analyte or Component Names 4-chloro-3-sulfamoyl-N-(2-methyl-1H-indol-1-yl)benzamide
Shipping Condition Room Temperature
Molecular Formula C16 H14 Cl N3 O3 S
Certifications/Compliance ISO 9001
Synonym Indapamide Imp. B (EP), Indapamide USP Related Compound A, Indapamide USP RC A, 4-Chloro-N-(2-methyl-1H-indol-1-yl)-3-sulphamoylbenzamide, Indapamide Related Compound A, Indapamide Impurity B
InChI Formula InChI=1S/C16H14ClN3O3S/c1-10-8-11-4-2-3-5-14(11)20(10)19-16(21)12-6-7-13(17)15(9-12)24(18,22)23/h2-9H,1H3,(H,19,21)(H2,18,22,23)
SMILES Cc1cc2ccccc2n1NC(=O)c3ccc(Cl)c(c3)S(=O)(=O)N
IUPAC Name 4-chloro-N-(2-methylindol-1-yl)-3-sulfamoylbenzamide
Molecular Weight (g/mol) 363.82
Quantity 25 mg
Formula Weight 363.0444 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.