missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Chloro-N-(4-fluorobenzyl)-3-sulfamoylbenzamide, 95%

Brand:  Alfa Aesar™ H65419.03

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15439636

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Specifications

Chemical Identifiers

MFCD08522711

Specifications

4-Chloro-N-(4-fluorobenzyl)-3-sulfamoylbenzamide
1g
342.77
95%
White
MFCD08522711
Solid
Videos
Documents