Learn More
4'-Chloroacetophenone-d7, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | 4'-Chloroacetophenone-D7 |
| CAS | 1174565-85-5 |
| Molecular Formula | C8 D7 Cl O |
| Quantity | 0.5 g |
| Synonym | Ethanone-2,2,2-d3, 1-(4-chlorophenyl-2,3,5,6-d4)- (ACI), 1-(4-Chloro-2,3,5,6-tetradeuteriophenyl)-2,2,2-trideuterioethanone, 1-(4-Chlorophenyl-2,3,5,6-d4)ethanone-2,2,2-d3, 4'-Chloroacetophenone-d7, 4'-Chloroacetophenone-D7, 4'-Chloroacetophenone-2,2,2,2',3',5',6'-d7 |
| InChI Formula | InChI=1 S/C8H7ClO/c1-6(10)7-2-4-8(9)5-3-7/h2-5 H,1H3/i1D3,2 D,3 D,4 D,5 D |
| SMILES | [2 H]c1c([2 H])c(C(=O)C([2 H])([2 H])[2 H])c([2 H])c([2 H])c1Cl |
| IUPAC Name | 1-(4-chloro-2,3,5,6-tetradeuteriophenyl)-2,2,2-trideuterioethanone |
| Molecular Weight (g/mol) | 161.6367 |
| Formula Weight | 161.0625 g/mol |
| Show More |