Learn More
4-Chloroaniline-2,3,5,6-d4, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | 4-Chloroaniline-d4 |
| CAS | 191656-33-4 |
| Molecular Formula | C6 D4 H2 Cl N |
| Quantity | 0.25 g |
| Synonym | Benzen-2,3,5,6-d4-amine, 4-chloro- (9 CI), 4-Chlorobenzen-2,3,5,6-d4-amine, 4-Chloroaniline-d4, 4-Chloroaniline-2,3,5,6-D4 |
| InChI Formula | InChI=1 S/C6H6ClN/c7-5-1-3-6(8)4-2-5/h1-4 H,8H2/i1D,2 D,3 D,4 D |
| SMILES | [2 H]c1c([2 H])c(Cl)c([2 H])c([2 H])c1N |
| IUPAC Name | 4-chloro-2,3,5,6-tetradeuterioaniline |
| Molecular Weight (g/mol) | 131.5962 |
| Formula Weight | 131.044 g/mol |
| Show More |