Learn More
4-Chlorobenzyl Cyanide, TRC
Product Code. 30137280
Change view
Click to view available options
Quantity:
10 g
100 g
Unit Size:
100g
10g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137280 100 g 100g
30012724 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30137280 Supplier Toronto Research Chemicals Supplier No. TRCC364720100G
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 140-53-4
Molecular Formula C8 H6 Cl N
Molecular Weight (g/mol) 151.59
Synonym (4-Chlorophenyl)acetonitrile, Sibutramine Hydrochloride Monohydrate Imp. E (Pharmeuropa), Benzeneacetonitrile, 4-chloro- (9CI, ACI), 4-Chlorobenzeneacetonitrile (ACI), Acetonitrile, (p-chlorophenyl)- (7CI, 8CI), (p-Chlorophenyl)acetonitrile, 2-(4-Chlorophenyl)acetonitrile, 4-Chlorobenzyl cyanide, 4-Chlorobenzyl nitrile, NSC 49108, p-Chlorbenzylnitrile, p-Chloro-α-cyanotoluene, p-Chlorobenzyl cyanide
IUPAC Name 2-(4-chlorophenyl)acetonitrile
SMILES Clc1ccc(CC#N)cc1
Specifications
Quantity 100 g
InChI Formula InChI=1S/C8H6ClN/c9-8-3-1-7(2-4-8)5-6-10/h1-4H,5H2
Formula Weight 151.0189
Chemical Name or Material (4-Chlorophenyl)acetonitrile