missing translation for 'onlineSavingsMsg'
Learn More

4-Chloronicotinic acid, 96%, Thermo Scientific™

Catalog Number p-4813107
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
10177-29-4
C6H4ClNO2
157.55
MFCD00128860
IMRGVWZLCZERSQ-UHFFFAOYSA-N
4-chloronicotinic acid, 4-chloronicotinicacid, 4-chloro-3-pyridinecarboxylic acid, 4-chloro-nicotinic acid, 3-pyridinecarboxylic acid, 4-chloro, 3-carboxy-4-chloropyridine, 4-chlornicotinsaure, pubchem12989, acmc-1br3v, 4-chloropyridine-3-carboxyl
818229
4-chloropyridine-3-carboxylic acid
OC(=O)C1=C(Cl)C=CN=C1
This item is not returnable. View return policy
10177-29-4
C6H4ClNO2
157.55
MFCD00128860
IMRGVWZLCZERSQ-UHFFFAOYSA-N
4-chloronicotinic acid, 4-chloronicotinicacid, 4-chloro-3-pyridinecarboxylic acid, 4-chloro-nicotinic acid, 3-pyridinecarboxylic acid, 4-chloro, 3-carboxy-4-chloropyridine, 4-chlornicotinsaure, pubchem12989, acmc-1br3v, 4-chloropyridine-3-carboxyl
818229
4-chloropyridine-3-carboxylic acid
OC(=O)C1=C(Cl)C=CN=C1
Melting Point 170°C to 171°C
Assay Percent Range 95% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 157.56
Percent Purity 96%
Chemical Name or Material 4-Chloronicotinic acid
compliance-icons
Product Identifier
  • 4-Chloronicotinic acid, 96%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'