Learn More
(4-Chlorophenyl)acetonitrile, Mikromol™
Description
(4-Chlorophenyl)acetonitrile
Specifications
Specifications
| Chemical Name or Material | (4-Chlorophenyl)acetonitrile |
| CAS | 140-53-4 |
| Analyte or Component Names | (4-Chlorophenyl)acetonitrile |
| Shipping Condition | Room Temperature |
| Molecular Formula | C8 H6 Cl N |
| Certifications/Compliance | ISO 9001 |
| Synonym | (4-Chlorophenyl)acetonitrile, Sibutramine Hydrochloride Monohydrate Imp. E (Pharmeuropa), Pyrimethamine Impurity C, Sibutramine Hydrochloride Monohydrate Impurity E, Sibutramine Impurity E |
| InChI Formula | InChI=1S/C8H6ClN/c9-8-3-1-7(2-4-8)5-6-10/h1-4H,5H2 |
| SMILES | Clc1ccc(CC#N)cc1 |
| IUPAC Name | 2-(4-chlorophenyl)acetonitrile |
| Show More |