missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorophenylboronic acid pinacol ester, 97%, ACROS Organics™

Brand:  Acros Organics 444690250

Additional Details : CAS : 4187-56-8 Weight : 0.07500kg


Quantity 25g

 View more versions of this product

Product Code. 11906071

  • € 198.00 / 25g


Estimated Shipment: 07-07-2021
Add to basket

Description

Description

Specifications

Chemical Identifiers

195062-61-4
238.52
NYARTXMDWRAVIX-UHFFFAOYSA-N
10633712
CC1(C)OB(OC1(C)C)C1=CC=C(Cl)C=C1
C12H16BClO2
MFCD05663875
2-4-chlorophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-chlorophenylboronic acid pinacol ester, 4-chlorophenylboronic acid, pinacol ester, 1,3,2-dioxaborolane,2-4-chlorophenyl-4,4,5,5-tetramethyl, 1,3,2-dioxaborolane, 2-4-chlorophenyl-4,4,5,5-tetramethyl, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl chlorobenzene, 4-chlorophenyl boronic acid pinacol ester, 4-chlorobenzeneboronic acid, pinacol ester, p-chlorophenylboronic acid 2,3-dimethyl-2,3-butanediyl
2-(4-chlorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Specifications

4-Chlorophenylboronic acid pinacol ester
55.0°C to 56.0°C
195062-61-4
C12H16BClO2
2-4-chlorophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-chlorophenylboronic acid pinacol ester, 4-chlorophenylboronic acid, pinacol ester, 1,3,2-dioxaborolane,2-4-chlorophenyl-4,4,5,5-tetramethyl, 1,3,2-dioxaborolane, 2-4-chlorophenyl-4,4,5,5-tetramethyl, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl chlorobenzene, 4-chlorophenyl boronic acid pinacol ester, 4-chlorobenzeneboronic acid, pinacol ester, p-chlorophenylboronic acid 2,3-dimethyl-2,3-butanediyl
CC1(C)OB(OC1(C)C)C1=CC=C(Cl)C=C1
238.52
238.52
>110°C
25g
96% min. (GC)
MFCD05663875
NYARTXMDWRAVIX-UHFFFAOYSA-N
2-(4-chlorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
10633712
97%
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 4-Chlorophenylboronic acid pinacol ester

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS Documents
Certificates