Learn More
4-Cyano-1-butyne, 97%, Thermo Scientific Chemicals
Catalog Number p-7077565
Click to view available options
Quantity:
1 g
5 g
19596-07-7
C5H5N
79.1
VMUWIDHKAIGONP-UHFFFAOYSA-N
4-cyano-1-butyne, 4-pentynenitrile, 1-cyano-3-butyne
140560
pent-4-ynenitrile
C#CCCC#N
19596-07-7
C5H5N
79.1
VMUWIDHKAIGONP-UHFFFAOYSA-N
4-cyano-1-butyne, 4-pentynenitrile, 1-cyano-3-butyne
140560
pent-4-ynenitrile
C#CCCC#N
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Density 0.9063g/mL
Boiling Point 79.0°C (39.0 mmHg)
Flash Point 65.5°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Linear Formula NCCH2CH2CCH
Quantity 1 g
Specific Gravity 0.9063
Formula Weight 79.1
Percent Purity 97%
Chemical Name or Material 4-Cyano-1-butyne
Show More Show Less
compliance-icons
Product Identifier
  • 4-Cyano-1-butyne
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.