missing translation for 'onlineSavingsMsg'
Learn More

4-Cyano-2-fluorobenzeneboronic acid, 98%, Thermo Scientific Chemicals

Catalog Number 15447638
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1150114-77-4
C7H5BFNO2
164.93
MFCD07644472
HFAUMAIVCYTVQR-UHFFFAOYSA-N
4-cyano-2-fluorophenyl boronic acid, boronic acid, b-4-cyano-2-fluorophenyl, acmc-2099nb, 4-borono-3-fluorobenzonitrile, 4-cyano-2-fluoro-phenylboronic acid, 4-cyano-2-fluorobenzeneboronic acid
44755209
(4-cyano-2-fluorophenyl)boronic acid
OB(O)C1=C(F)C=C(C=C1)C#N
This item is not returnable. View return policy
1150114-77-4
C7H5BFNO2
164.93
MFCD07644472
HFAUMAIVCYTVQR-UHFFFAOYSA-N
4-cyano-2-fluorophenyl boronic acid, boronic acid, b-4-cyano-2-fluorophenyl, acmc-2099nb, 4-borono-3-fluorobenzonitrile, 4-cyano-2-fluoro-phenylboronic acid, 4-cyano-2-fluorobenzeneboronic acid
44755209
(4-cyano-2-fluorophenyl)boronic acid
OB(O)C1=C(F)C=C(C=C1)C#N

4-Cyano-2-fluorobenzeneboronic acid is used an active pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Cyano-2-fluorobenzeneboronic acid is used an active pharmaceutical intermediate.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 164.93
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 4-Cyano-2-fluorobenzeneboronic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'