missing translation for 'onlineSavingsMsg'
Learn More

4-Cyano-D-phenylalanine, 98%, Thermo Scientific™

Catalog Number p-7055173
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
263396-44-7
C10H10N2O2
190.202
MFCD00270364
KWIPUXXIFQQMKN-SECBINFHSA-N
r-2-amino-3-4-cyanophenyl propanoic acid, d-4-cyanophenylalanine, h-d-phe 4-cn-oh, 4-cyano-d-phenylalanine, 2r-2-amino-3-4-cyanophenyl propanoic acid, d-4-cn-phe-oh, d-4-cyanophe, h-4-cyano-phe-oh, ambotzhaa7570, pubchem13160
1501864
(2R)-2-amino-3-(4-cyanophenyl)propanoic acid
C1=CC(=CC=C1CC(C(=O)O)N)C#N
This item is not returnable. View return policy
263396-44-7
C10H10N2O2
190.202
MFCD00270364
KWIPUXXIFQQMKN-SECBINFHSA-N
r-2-amino-3-4-cyanophenyl propanoic acid, d-4-cyanophenylalanine, h-d-phe 4-cn-oh, 4-cyano-d-phenylalanine, 2r-2-amino-3-4-cyanophenyl propanoic acid, d-4-cn-phe-oh, d-4-cyanophe, h-4-cyano-phe-oh, ambotzhaa7570, pubchem13160
1501864
(2R)-2-amino-3-(4-cyanophenyl)propanoic acid
C1=CC(=CC=C1CC(C(=O)O)N)C#N

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
4-Cyano-D-phenylalanine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Store in freezer at(-20°C). Keep container tightly closed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Color White
UN Number UN3439
Solubility Information Slightly soluble in water.
Formula Weight 190.2
Percent Purity 98%
Quantity 250 mg
Physical Form Solid
Chemical Name or Material 4-Cyano-D-phenylalanine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'