Learn More
4-Cyanocyclohexanone, TRC
Specifications
Specifications
| Chemical Name or Material | 4-Cyanocyclohexanone |
| CAS | 34916-10-4 |
| Molecular Formula | C7H9NO |
| Quantity | 500 mg |
| Synonym | 4-Oxocyclohexanecarbonitrile |
| InChI Formula | InChI=1S/C7H9NO/c8-5-6-1-3-7(9)4-2-6/h6H,1-4H2 |
| SMILES | N#CC(CC1)CCC1=O |
| IUPAC Name | 4-oxocyclohexane-1-carbonitrile |
| Molecular Weight (g/mol) | 123.15 |
| Formula Weight | 123.07 |