Learn More
4-Cyanophenylacetonitrile, 97%
Product Code. 15460498
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15460498 5 g 5g
15450498 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15460498 Supplier Thermo Scientific Alfa Aesar Supplier No. H54112.06
Request Bulk or Custom Format
CAS: 876-31-3 | C9H6N2 | 142.16 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 876-31-3
Molecular Formula C9H6N2
Molecular Weight (g/mol) 142.16
MDL Number MFCD00060305
InChI Key QILKKAFYAFEWGU-UHFFFAOYSA-N
Synonym 4-cyanomethyl benzonitrile, 4-cyanophenylacetonitrile, benzeneacetonitrile, 4-cyano, p-tolunitrile, .alpha.-cyano, p-cyanobenzylcyanide, 4-cyanobenzeneacetonitrile, .alpha.-cyano-p-tolunitrile, 4-cyanbenzylcyanide, 4-cyanobenzylcyanide, acmc-209qoz
PubChem CID 136686
IUPAC Name 4-(cyanomethyl)benzonitrile
SMILES N#CCC1=CC=C(C=C1)C#N
Specifications
Density 1.056
Boiling Point 180°C to 181°C
Flash Point 71°C (160°F)
Refractive Index 1.499
Quantity 5 g
Formula Weight 142.16
Percent Purity 97%
Chemical Name or Material 4-Cyanophenylacetonitrile