Learn More
4-(Diethylphosphono)-3-methyl-2-butenenitrile, E/Z mixture, TRC
Specifications
Specifications
| Chemical Name or Material | 4-(Diethylphosphono)-3-methyl-2-butenenitrile |
| CAS | 87549-50-6 |
| Molecular Formula | C9 H16 N O3 P |
| Quantity | 500 mg |
| InChI Formula | InChI=1S/C9H16NO3P/c1-4-12-14(11,13-5-2)8-9(3)6-7-10/h6H,4-5,8H2,1-3H3/b9-6+ |
| SMILES | CCOP(=O)(C\C(=C\C#N)\C)OCC |
| IUPAC Name | (E)-4-diethoxyphosphoryl-3-methylbut-2-enenitrile |
| Molecular Weight (g/mol) | 217.2 |
| Formula Weight | 217.0868 |