Learn More
4-[(E)-2-Phenylethenyl]pyridine, TRC
Specifications
Specifications
| Chemical Name or Material | 4-[(E)-2-Phenylethenyl]pyridine |
| CAS | 5097-93-8 |
| Molecular Formula | C13 H11 N |
| Quantity | 500 mg |
| Synonym | 4-[(1E)-2-Phenylethenyl]pyridine, (E)-4-(2-Phenylethenyl)pyridine, (E)-4-Styrylpyridine, (E)-4-Stilbazole, 4-trans-Styrylpyridine, trans-4-Stilbazole, trans-4-Styrylpyridine |
| InChI Formula | InChI=1S/C13H11N/c1-2-4-12(5-3-1)6-7-13-8-10-14-11-9-13/h1-11H/b7-6+ |
| SMILES | C(=C\c1ccncc1)/c2ccccc2 |
| IUPAC Name | 4-[(E)-2-phenylethenyl]pyridine |
| Molecular Weight (g/mol) | 181.233 |
| Formula Weight | 181.089 |