Learn More
4-(Ethylthio)benzaldehyde, 97+%
Product Code. 11398566
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11398566 1 g 1g
11308576 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11398566 Supplier Thermo Scientific Alfa Aesar Supplier No. L17624.03
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
missing translation for 'validMainframeMsg'
CAS: 84211-94-9 | C9H10OS | 166.238 g/mol
It is an active pharmaceutical ingredient.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is an active pharmaceutical ingredient.
Solubility
Difficult to mix in water.
Notes
Air sensitive. Store in cool dry conditions in well sealed containers. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 84211-94-9
Molecular Formula C9H10OS
Molecular Weight (g/mol) 166.238
MDL Number MFCD01318145
InChI Key LPRKZLUVZOFDDT-UHFFFAOYSA-N
Synonym 4-ethylthio benzaldehyde, 4-ethylsulfanyl benzaldehyde, 4-ethylthiobenzaldehyde, 4-ethylsulfanyl-benzaldehyde, benzaldehyde,4-ethylthio
PubChem CID 7006654
IUPAC Name 4-ethylsulfanylbenzaldehyde
SMILES CCSC1=CC=C(C=C1)C=O
Specifications
Boiling Point 151°C to 153°C (15 mmHg)
Refractive Index 1.624
Quantity 1 g
Beilstein 1863857
Sensitivity Air Sensitive
Solubility Information Difficult to mix in water.
Formula Weight 166.24
Percent Purity 98%
Chemical Name or Material 4-(Ethylthio)benzaldehyde