Learn More
4-Fluoro-2-iodoaniline, TRC
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | 4-Fluoro-2-iodoaniline |
| CAS | 61272-76-2 |
| Molecular Formula | C6H5FIN |
| Quantity | 5 g |
| InChI Formula | InChI=1S/C6H5FIN/c7-4-1-2-6(9)5(8)3-4/h1-3H,9H2 |
| SMILES | Nc1ccc(F)cc1I |
| Molecular Weight (g/mol) | 237.01 |
| Formula Weight | 236.9451 |