missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-2-methoxybenzylamine, 97%

Product Code. p-7029343
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
11356899 1 g 1g
11366899 5 g 5g
2 options
This item is not returnable. View return policy

Product Code. 11356899

Brand: Thermo Scientific Alfa Aesar H26796.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 870563-60-3 | C8H10FNO | 155.17 g/mol

As pharmaceutical intermediate. As a metabolite of Capsaicin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
As pharmaceutical intermediate. As a metabolite of Capsaicin.

Solubility
Insoluble in water.

Notes
Air Sensitive and Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 870563-60-3
Molecular Formula C8H10FNO
Molecular Weight (g/mol) 155.17
MDL Number MFCD04116361
InChI Key OUYQGZMPYJPPER-UHFFFAOYSA-N
Synonym 4-fluoro-2-methoxyphenyl methanamine, 4-fluoro-2-methoxybenzylamine, 2-methoxy-4-fluorobenzylamine, benzenemethanamine, 4-fluoro-2-methoxy, 1-4-fluoro-2-methoxyphenyl methanamine, 4-fluoro-2-methoxy-benzylamine
PubChem CID 17750694
IUPAC Name (4-fluoro-2-methoxyphenyl)methanamine
SMILES COC1=C(CN)C=CC(F)=C1

Specifications

Refractive Index 1.524
Quantity 1 g
UN Number UN2735
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 155.17
Percent Purity 97%
Chemical Name or Material 4-Fluoro-2-methoxybenzylamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.