missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-3-formylbenzeneboronic acid, 98+%, Thermo Scientific™

Product Code. 15499069
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15499069

Brand: Thermo Scientific Alfa Aesar L17808.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 374538-01-9
Molecular Formula C7H6BFO3
Molecular Weight (g/mol) 167.93
MDL Number MFCD02093074
InChI Key YABSTJQEBSKPCG-UHFFFAOYSA-N
Synonym 4-fluoro-3-formylbenzeneboronic acid, 4-fluoro-3-formylphenyl boronic acid, boronic acid, 4-fluoro-3-formylphenyl, 4-fluoro-3-formyl-phenyl boronic acid, pubchem10809, acmc-1cswy, 3-formyl 4-fluorophenyl boronic acid, 5-borono-2-fluorobenzaldehyde, 3-formyl-4-fluorophenyl boronic acid, 4-fluoro-3-formyl benzene boronic acid
PubChem CID 2778651
IUPAC Name (4-fluoro-3-formylphenyl)boronic acid
SMILES B(C1=CC(=C(C=C1)F)C=O)(O)O

Specifications

Melting Point ∼240°C
Quantity 5 g
Beilstein 9046762
Sensitivity Air Sensitive
Formula Weight 167.93
Percent Purity ≥98%
Chemical Name or Material 4-Fluoro-3-formylbenzeneboronic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.