missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-3-iodobenzoic acid, 97%, Thermo Scientific™

Product Code. 15476617
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15476617 250 mg 250mg
15466617 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15476617 Supplier Thermo Scientific Alfa Aesar Supplier No. H33762.MD
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 403-18-9
Molecular Formula C7H4FIO2
Molecular Weight (g/mol) 266.01
MDL Number MFCD03788556
InChI Key IKSOHJRYPQRTLQ-UHFFFAOYSA-N
Synonym benzoic acid, 4-fluoro-3-iodo, 3-iodo-4-fluorobenzoic acid, 4-fluoro-3-iodo-benzoic acid, pubchem1390, buttpark 122, buttpark 122\01-04
PubChem CID 10361
IUPAC Name 4-fluoro-3-iodobenzoic acid
SMILES C1=CC(=C(C=C1C(=O)O)I)F

Specifications

Melting Point 173°C to 175°C
Quantity 250 mg
Sensitivity Light sensitive
Formula Weight 266.01
Percent Purity 97%
Chemical Name or Material 4-Fluoro-3-iodobenzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.