Learn More
4-Fluoro-beta-nitrostyrene, 98%, Thermo Scientific™
Catalog Number p-7055904
Click to view available options
Quantity:
5 g
25 g
100 g
706-08-1
C8H6FNO2
167.14
MFCD00024824
VRFSQVFSQAYHRU-AATRIKPKSA-N
1-fluoro-4-2-nitrovinyl benzene, 4-fluoro-beta-nitrostyrene, e-1-fluoro-4-2-nitrovinyl benzene, trans-p-fluoro-beta-nitrostyrene, p-fluoro-beta-nitrostyrene, styrene, p-fluoro-beta-nitro, 1-4-fluorophenyl-2-nitroethylene, trans p-fluorophenyl-1 nitro-2 ethylene french, styrene, p-fluoro-beta-nitro-, e, 1-fluoro-4-e-2-nitrovinyl benzene
5381680
[O-][N+](=O)\C=C\C1=CC=C(F)C=C1
706-08-1
C8H6FNO2
167.14
MFCD00024824
VRFSQVFSQAYHRU-AATRIKPKSA-N
1-fluoro-4-2-nitrovinyl benzene, 4-fluoro-beta-nitrostyrene, e-1-fluoro-4-2-nitrovinyl benzene, trans-p-fluoro-beta-nitrostyrene, p-fluoro-beta-nitrostyrene, styrene, p-fluoro-beta-nitro, 1-4-fluorophenyl-2-nitroethylene, trans p-fluorophenyl-1 nitro-2 ethylene french, styrene, p-fluoro-beta-nitro-, e, 1-fluoro-4-e-2-nitrovinyl benzene
5381680
[O-][N+](=O)\C=C\C1=CC=C(F)C=C1
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
It is used as pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Store at 99-102°C. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 99°C to 102°C
Color Yellow
Quantity 5 g
UN Number UN2811
Beilstein 2208372
Solubility Information Slightly soluble in water.
IUPAC Name 1-fluoro-4-[(1E)-2-nitroethenyl]benzene
Formula Weight 167.14
Percent Purity 98%
Physical Form Crystalline Solid
Chemical Name or Material 4-Fluoro-beta-nitrostyrene